Oxoid™ Post-Pasteurization Contamination Test Selective Supplement
Product Image
Thermo Scientific™
Oxoid™ Post-Pasteurization Contamination Test Selective Supplement
The Oxoid Post-Pasteurization Contamination Test Selective Supplement (PPCT Supplement) is used in the dairy industry.
Have Questions?
Catalog NumberQuantity
SR0159E10/Pk.
Catalog number SR0159E
Price (USD)
-
Request A Quote
Quantity:
10/Pk.
Ask our AI about this Product
Oxoid Post-Pasteurization Contamination Test Selective Supplement (PPCT Supplement) is used in the dairy industry.
  • Add to Milk Agar, Part No. CM0021B
  • Each vial supplements 200mL of medium
Specifications
DescriptionPost-Pasteurization Contamination Test Selective Supplement
FormFreeze-dried
Product TypeSelective Supplement
Quantity10/Pk.
YieldEach vial for 200mL medium
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
756200-COACertificate of AnalysisJul 21, 2017SR0159E
778335-COACertificate of AnalysisJul 21, 2017SR0159E
695927-COACertificate of AnalysisJul 21, 2017SR0159E
3 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection