1x1 image pixel for data collection
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors with clamps, injection site
Please note: order placement quantity must be in multiples of 25, minimum of 25 required.
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors with clamps, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors with clamps, injection site

The 10 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Catalog NumberQuantity
SS00017I
also known as SS00017-I
25/Cs.
Catalog number SS00017I
also known as SS00017-I
Price (USD)
-
Request A Quote
Quantity:
25/Cs.
The 10 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (1 MPC insert + cap and pinch clamp, 1 MPC body + cap and pinch clamp, 1 injection site).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Dimensions (L x W x H)56.6 x 32.5 cm (22.3 x 12.8 in.)
Film DescriptionASI™ 28
Includes3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Outer PackagingOne 4-mil. poly bag
Port DetailMPC insert and body connectors, plug and injection site
Product LineLabtainer™
Quantity25/Cs.
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Tubing StyleEVA
Capacity (Metric)10 L
Length (Metric)56.6 cm
MaterialEVA
Product TypeBioprocess Container
SterilitySterile
Width (English)12.8 in.
Width (Metric)32.5 cm
Unit Size10 L
Contents & Storage
Store at room temperature.

Share catalog number, name or link