Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A214 pH and ISE Benchtop Meter. This meter is ideal for a wide range of applications and advanced pH or ion analysis in the lab. Perform up to 5 point pH and ISE calibrations, log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) |
Outputs | USB, RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | High/low limit alarm, calibration due alarm |
Backlight Option | Yes, selectable |
Battery Life | 800 hr. |
Battery Type | 4 AA (sold separately) |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 5 |
Calibration Points (pH) | 1 to 5 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Star A214 pH/ISE benchtop meter kit with fluoride electrode and solutions, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe and stand |
Channels | 1 |
Display Type | LCD Graphic, Backlit |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 3 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 9609BNWP Fluoride Electrode; 096019 Stirrer Probe; 040906 Fluoride 1ppm with TISAB II Standard; 040907 2ppm with TISAB II Standard; 040908 Fluoride 10ppm with TISAB II Standard; 940909 TISAB II Solution; Electrode Arm with redesigned holder; Universal Power Adapter |
Inputs | BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) |
Isopotential Point | 7.000pH |
Keypad | Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update |
Log Function Type | Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel with password protection |
Model | Orion Star A214 pH/ISE/mV/RmV/ORP/Temperature Benchtop Meter |
Power Supply | Universal AC adapter (included) or 4 AA batteries (sold separately) |
Range (ISE) | 0.0001 to 19900 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 Year Meter |
Dimensions (L x W x H) | 24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.) |
Weight (English) | 2 lb. |
Weight (Metric) | 0.9 kg |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) |
Outputs | USB, RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | High/low limit alarm, calibration due alarm |
Backlight Option | Yes, selectable |
Battery Life | 800 hr. |
Battery Type | 4 AA (sold separately) |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 5 |
Calibration Points (pH) | 1 to 5 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Star A214 pH/ISE benchtop meter with stand |
Channels | 1 |
Display Type | LCD Graphic, Backlit |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 3 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Star A214 pH/ISE meter, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
Inputs | BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) |
Isopotential Point | 7.000pH |
Keypad | Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update |
Log Function Type | Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel with password protection |
Model | Orion Star A214 pH/ISE/mV/RmV/ORP/Temperature Benchtop Meter |
Power Supply | Universal AC adapter (included) or 4 AA batteries (sold separately) |
Probe Type | pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (purchase separately) |
Range (ISE) | 0.0001 to 19900 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 Year Meter |
Dimensions (L x W x H) | 24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.) |
Weight (English) | 2 lb. |
Weight (Metric) | 0.9 kg |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) |
Outputs | USB, RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | High/low limit alarm, calibration due alarm |
Backlight Option | Yes, selectable |
Battery Life | 800 hr. |
Battery Type | 4 AA (sold separately) |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 5 |
Calibration Points (pH) | 1 to 5 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Star A214 pH/ISE benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and stand |
Channels | 1 |
Display Type | LCD Graphic, Backlit |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 3 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 096019 Stirrer Probe; 810199 ROSS pH Buffer Kit |
Inputs | BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) |
Isopotential Point | 7.000pH |
Keypad | Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update |
Log Function Type | Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel with password protection |
Model | Orion Star A214 pH/ISE/mV/RmV/ORP/Temperature Benchtop Meter |
Power Supply | Universal AC adapter (included) or 4 AA batteries (sold separately) |
Probe Type | pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8102BNUWP ROSS Ultra pH electrode, 927007MD Orion ATC probe, 096019 Orion Star stirrer probe) |
Range (ISE) | 0.0001 to 19900 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 Year Meter |
Dimensions (L x W x H) | 24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.) |
Weight (English) | 2 lb. |
Weight (Metric) | 0.9 kg |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) |
Outputs | USB, RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | High/low limit alarm, calibration due alarm |
Backlight Option | Yes, selectable |
Battery Life | 800 hr. |
Battery Type | 4 AA (sold separately) |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 5 |
Calibration Points (pH) | 1 to 5 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Star A214 pH/ISE benchtop meter kit with ammonia electrode and solutions, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe and stand |
Channels | 1 |
Display Type | LCD Graphic, Backlit |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 3 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 9512HPBNWP High-Performance Ammonia Electrode; 096019 Stirrer Probe; 951007 Ammonia 1000ppm Standard; 951210 Low-level Ammonia ISA; 951213 Ammonia ISE Storage Solution; 951213 Ammonia ISE Storage Solution; Electrode Arm with Holder |
Inputs | BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) |
Isopotential Point | 7.000pH |
Keypad | Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update |
Log Function Type | Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel with password protection |
Model | Orion Star A214 pH/ISE/mV/RmV/ORP/Temperature Benchtop Meter |
Power Supply | Universal AC adapter (included) or 4 AA batteries (sold separately) |
Range (ISE) | 0.0001 to 19900 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 Year Meter |
Dimensions (L x W x H) | 24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.) |
Weight (English) | 2 lb. |
Weight (Metric) | 0.9 kg |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) |
Outputs | USB, RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | High/low limit alarm, calibration due alarm |
Backlight Option | Yes, selectable |
Battery Life | 800 hr. |
Battery Type | 4 AA (sold separately) |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 5 |
Calibration Points (pH) | 1 to 5 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Star A214 pH/ISE benchtop meter kit with ROSS sodium electrode and solutions, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe and stand |
Channels | 1 |
Display Type | LCD Graphic, Backlit |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 3 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 8611BNWP ROSS Sodium Electrode with Sodium Standards, Reagents and Solutions; 096019 Stirrer Probe; Electrode Arm with Redesigned Holder; Universal Power Adapter |
Inputs | BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) |
Isopotential Point | 7.000pH |
Keypad | Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update |
Log Function Type | Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel with password protection |
Model | Orion Star A214 pH/ISE/mV/RmV/ORP/Temperature Benchtop Meter |
Power Supply | Universal AC adapter (included) or 4 AA batteries (sold separately) |
Range (ISE) | 0.0001 to 19900 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 Year Meter |
Dimensions (L x W x H) | 24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.) |
Weight (English) | 2 lb. |
Weight (Metric) | 0.9 kg |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Includes | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
STARA2147 | Each | Orion Star A214 pH/ISE benchtop meter kit with fluoride electrode and solutions, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe and stand | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 9609BNWP Fluoride Electrode; 096019 Stirrer Probe; 040906 Fluoride 1ppm with TISAB II Standard; 040907 2ppm with TISAB II Standard; 040908 Fluoride 10ppm with TISAB II Standard; 940909 TISAB II Solution; Electrode Arm with redesigned holder; Universal Power Adapter | 3,177.00 Online price 3,530.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
STARA2140 | Each | Orion Star A214 pH/ISE benchtop meter with stand | Star A214 pH/ISE meter, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate | 1,584.00 Online price 1,760.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
STARA2145 | Each | Orion Star A214 pH/ISE benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and stand | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 096019 Stirrer Probe; 810199 ROSS pH Buffer Kit | 2,205.00 Online price 2,450.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
STARA2146 | Each | Orion Star A214 pH/ISE benchtop meter kit with ammonia electrode and solutions, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe and stand | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 9512HPBNWP High-Performance Ammonia Electrode; 096019 Stirrer Probe; 951007 Ammonia 1000ppm Standard; 951210 Low-level Ammonia ISA; 951213 Ammonia ISE Storage Solution; 951213 Ammonia ISE Storage Solution; Electrode Arm with Holder | 2,961.00 Online price 3,290.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
STARA2148 | Each | Orion Star A214 pH/ISE benchtop meter kit with ROSS sodium electrode and solutions, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe and stand | Star A214 pH/ISE Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 8611BNWP ROSS Sodium Electrode with Sodium Standards, Reagents and Solutions; 096019 Stirrer Probe; Electrode Arm with Redesigned Holder; Universal Power Adapter | 2,736.00 Online price 3,040.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific Orion Star A214 pH/ISE benchtop meters are easy to use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.
Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.