Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A325 pH/Conductivity Portable Multiparameter Meter. This meter is designed for a wide range of pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Use the two channels to measure pH and conductivity simultaneously or view each channel separately. Log up to 5000 data point sets with time/date stamp and transfer logs to a printer or computer.
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003) |
Outputs | USB, RS232 |
Accuracy (Conductivity) | 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Accuracy (Resistivity) | 0.5% of reading ±1 digit |
Accuracy (Salinity) | 0.5% of reading ±1 digit |
Accuracy (TDS) | 0.5% of reading ±1 digit |
Alarm Output | High/low limit alarm, calibration due alarm |
Backlight Option | Yes, selectable |
Battery Life | 800 hr. |
Battery Type | 4 AA (included) |
Calibration | pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (Conductivity) | Up to 5 |
Calibration Points (pH) | 1 to 5 |
Cell Constant | 0.001 to 199.9cm-1 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 5000 with date and time stamp |
Description | Orion Star A325 pH/conductivity portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, DuraProbe 4-cell conductivity sensor, solutions, meter armor and field case |
Channels | 2 |
Display Type | LCD Graphic, Backlit |
IP Rating | IP 67 |
Measurement Modes | pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature |
Range (Conductivity) | 0.001 μS/cm to 3000 mS/cm |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Range (Resistivity) | 2 ohm to 100.0 meg-ohm |
Range (Salinity) | 0.06 to 80.00 psu, 0.05 to 42.00 ppt |
Range (TDS) | 0.001 ppm to 200.0 ppt |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Orion STAR A325 pH/conductivity portable meter; 8107UWMMD Orion ROSS Triode pH/ATC gel-filled electrode with 3m cable; 013010MD Orion DuraProbe conductivity cell with 3m cable; 910410 pH 4.01 buffer pouches, pack of 10; 910710 pH 7.00 buffer pouches, pack of 10; 911010 pH 10.01 buffer pouches, pack of 10; 01100710 conductivity standard pouches, 1413 uS; 911110 rinse solution, pack of 10; protective armor with electrode holder; hard carrying case and batteries |
Inputs | BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe) |
Isopotential Point | 7.000pH |
Keypad | Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update |
Log Function Type | Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel with password protection |
Model | Orion Star A325 pH/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter |
Power Supply | 4 AA batteries (included) or universal AC adapter (sold separately) |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (Conductivity) | 0.001 μS minimum, auto ranging up to 4 significant digits |
Resolution (mV) | 0.1mV |
Resolution (Resistivity) | 1 ohm or 0.1 meg-ohm, auto ranging |
Resolution (Salinity) | 0.01 psu or 0.01 ppt minimum, auto ranging |
Resolution (TDS) | 0.001 ppm minimum, auto ranging up to 4 significant digits |
Salinity Measurement Modes | Practical salinity (psu) or natural sea water (ppt) |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
TDS Factor Range | Linear (0.02 to 9.99) |
Temperature Accuracy | ±0.1°C |
Temperature Compensation | Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature (Metric) Reference | 5, 10, 15, 20, 25°C |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 Year Meter |
Dimensions (L x W x H) | 24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.) |
Weight (English) | 1.2 lb. |
Weight (Metric) | 0.5 kg |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003) |
Outputs | USB, RS232 |
Accuracy (Conductivity) | 0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Accuracy (Resistivity) | 0.5% of reading ±1 digit |
Accuracy (Salinity) | 0.5% of reading ±1 digit |
Accuracy (TDS) | 0.5% of reading ±1 digit |
Alarm Output | High/low limit alarm, calibration due alarm |
Backlight Option | Yes, selectable |
Battery Life | 800 hr. |
Battery Type | 4 AA (included) |
Calibration | pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (Conductivity) | Up to 5 |
Calibration Points (pH) | 1 to 5 |
Cell Constant | 0.001 to 199.9cm-1 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 5000 with date and time stamp |
Description | Orion Star A325 pH/conductivity portable meter |
Channels | 2 |
Display Type | LCD Graphic, Backlit |
IP Rating | IP 67 |
Measurement Modes | pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature |
Range (Conductivity) | 0.001 μS/cm to 3000 mS/cm |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Range (Resistivity) | 2 ohm to 100.0 meg-ohm |
Range (Salinity) | 0.06 to 80.00 psu, 0.05 to 42.00 ppt |
Range (TDS) | 0.001 ppm to 200.0 ppt |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Star A325 pH/conductivity meter, 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate |
Inputs | BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe) |
Isopotential Point | 7.000pH |
Keypad | Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update |
Log Function Type | Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel with password protection |
Model | Orion Star A325 pH/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter |
Power Supply | 4 AA batteries (included) or universal AC adapter (sold separately) |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (Conductivity) | 0.001 μS minimum, auto ranging up to 4 significant digits |
Resolution (mV) | 0.1mV |
Resolution (Resistivity) | 1 ohm or 0.1 meg-ohm, auto ranging |
Resolution (Salinity) | 0.01 psu or 0.01 ppt minimum, auto ranging |
Resolution (TDS) | 0.001 ppm minimum, auto ranging up to 4 significant digits |
Salinity Measurement Modes | Practical salinity (psu) or natural sea water (ppt) |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
TDS Factor Range | Linear (0.02 to 9.99) |
Temperature Accuracy | ±0.1°C |
Temperature Compensation | Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature (Metric) Reference | 5, 10, 15, 20, 25°C |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 Year Meter |
Dimensions (L x W x H) | 24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.) |
Weight (English) | 1.2 lb. |
Weight (Metric) | 0.5 kg |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Includes | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
STARA3255 | Each | Orion Star A325 pH/conductivity portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, DuraProbe 4-cell conductivity sensor, solutions, meter armor and field case | Orion STAR A325 pH/conductivity portable meter; 8107UWMMD Orion ROSS Triode pH/ATC gel-filled electrode with 3m cable; 013010MD Orion DuraProbe conductivity cell with 3m cable; 910410 pH 4.01 buffer pouches, pack of 10; 910710 pH 7.00 buffer pouches, pack of 10; 911010 pH 10.01 buffer pouches, pack of 10; 01100710 conductivity standard pouches, 1413 uS; 911110 rinse solution, pack of 10; protective armor with electrode holder; hard carrying case and batteries | 2,484.00 Online price 2,760.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
STARA3250 | Each | Orion Star A325 pH/conductivity portable meter | Star A325 pH/conductivity meter, 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate | 1,467.00 Online price 1,630.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.