Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Orion Star T900 series reagent bottle holder |
Type | Titrator accessory, reagent holder |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series burette cover |
Type | Titrator accessory, burette |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series 10ml burette |
Type | Titrator accessory, burette |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series 20ml burette |
Type | Titrator accessory, burette |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series 50ml burette |
Type | Titrator accessory, burette |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series 1L plastic bottle |
Type | Titrator accessory, reagent bottle |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series 1L glass amber bottle |
Type | Titrator accessory, reagent bottle |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series 1L plastic bottles, pack of 12 |
Type | Titrator accessory, reagent bottle |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series 1L glass amber bottles, pack of 12 |
Type | Titrator accessory, reagent bottle |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Description | Orion Star T900 series GL38 reagent bottle cap with ports for tubing and drying tube |
Type | Titrator accessory, reagent cap |
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
START-BT0 | Each | Orion Star T900 series reagent bottle holder | 49.95 Online price 55.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-B00 | Each | Orion Star T900 series burette cover | 49.95 Online price 55.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-B10 | Each | Orion Star T900 series 10ml burette | 433.80 Online price 482.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-B20 | Each | Orion Star T900 series 20ml burette | 433.80 Online price 482.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-B50 | Each | Orion Star T900 series 50ml burette | 433.80 Online price 482.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-BT1 | Each | Orion Star T900 series 1L plastic bottle | 27.90 Online price 31.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-BT2 | Each | Orion Star T900 series 1L glass amber bottle | 27.90 Online price 31.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-BT3 | Each | Orion Star T900 series 1L plastic bottles, pack of 12 | 192.60 Online price 214.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-BT4 | Each | Orion Star T900 series 1L glass amber bottles, pack of 12 | 192.60 Online price 214.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
START-CP1 | Each | Orion Star T900 series GL38 reagent bottle cap with ports for tubing and drying tube | 61.02 Online price 67.80 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|