HyPerforma™ 5:1 Single-Use Bioreactor, jacketed, AC motor, load cells
HyPerforma™ 5:1 Single-Use Bioreactor, jacketed, AC motor, load cells
Thermo Scientific™
HyPerforma™ 5:1 Single-Use Bioreactor, jacketed, AC motor, load cells
This 50 L Thermo Scientific Single-Use Bioreactor (S.U.B.), water-jacketed, with AC motor and load cells, is designed using traditional stainless-steelRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SUB00508100
also known as SUB0050.8100
50 L
SUB01008200
also known as SUB0100.8200
100 L
SUB02508300
also known as SUB0250.8300
250 L
SUB05008400
also known as SUB0500.8400
500 L
Catalog number SUB00508100
also known as SUB0050.8100
Price (USD)
-
Request A Quote
Capacity (Metric):
50 L
Ask our AI about this Product
This 50 L Thermo Scientific Single-Use Bioreactor (S.U.B.), water-jacketed, with AC motor and load cells, is designed using traditional stainless-steel bioreactor principles to ensure optimal cell culture performance. The S.U.B. is a fully functional bioreactor vessel including single-use sterile contact surfaces for mixing, venting, sparging, and temperature sensing. The complete line of S.U.B.s include 50, 100, 250, 500, 1000, and 2000 L sizes that ensure consistent scalability from pilot-scale studies, pre-clinical to commercial production.
Features:
• Unique proprietary mixing system that enables conventional overhead mixing while maintaining sterility and integrity of the S.U.B. BioProcess Container (BPC)
• Largest qualified standard component library, which allows for customization of S.U.B. BPCs
• Minimized vessel footprint and reduced hold-up volumes
• Offered with AC or DC motors designed for precise agitation control
• Optional cable management tree provides line set and cabling organization for ease of use
• Load cells offered as standard feature with analog or digital communication options
• Option to use industry-leading single-use and traditional sensors for pH, DO, and pressure monitoring
• Option to choose the bioreactor vessel as a stand-alone system or a complete turnkey system with dedicated controls from either the Finesse™ or Applikon™ families of control systems for bioproduction. These systems work on PC, Delta V™, Allen-Bradley™, or Siemens™ software platforms.
• Updated 5:1 mixing turn-down ratio in combination with patented cross-flow sparger technology introduces gas just above liquid’s surface—to facilitate efficient mass transfer CO2 stripping, and reduce bubble sparging and associated foam damage to cells. This feature allows customers to do more with fewer vessels—which improves productivity and reduces costs simultaneously.
Applications of the Thermo Scientific S.U.B. include:
• Batch, fed batch, and perfusion cultures
• Suspension and microcarrier cultures
• Proven for various cell lines such as CHO, Sp2/0 hybridoma, PER.C6, HEK293, Vero, MDCK, etc.
Specifications
Capacity (Metric)50 L
MaterialStainless Steel
Product LineHyPerforma™
Quantity50 L
Unit Size50 L
Certificates of Analysis
Search for Certificates of Analysis for your custom and standard Thermo Scientific™ single-use products.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link