The 200L Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum is part of a family of plastic tanks, drums and accessories for use with with 3-D bioprocess container systems and tank liners.
Features: • Includes lid and clamps • Available in cylindrical, conical and flat-bottom • 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port
Specifications
Capacity (Metric)
200 L
Description
Flat-bottom Outer Support Container for Bioprocess Bags, Port detail 4 and 1.75 in.