HyPerforma™ Smartainer™ Hardware, 200L, Stainless steel outer container
HyPerforma™ Smartainer™ Hardware, 200L, Stainless steel outer container
Thermo Scientific™
HyPerforma™ Smartainer™ Hardware, 200L, Stainless steel outer container
The 200L Thermo Scientific™ HyPerforma™ Smartainer is a stainless-steel non-jacketed outer container system designed to accommodate large volume bioprocess containers.Features:•Read more
Have Questions?
Catalog number SV50160.01
Price (USD)
-
Request A Quote
The 200L Thermo Scientific™ HyPerforma™ Smartainer is a stainless-steel non-jacketed outer container system designed to accommodate large volume bioprocess containers.
Features:
• Durable containers are constructed from 304 stainless steel
• Ideal for in-house handling applications in critical areas
• Movable and stackable with pallet jack or forklift
• Non-jacketed
• Allow both top and bottom discharge from BPCs
Specifications
Capacity (Metric)200 L
DimensionsInternal Dimensions (W x D x H): 73 cm x 53.3 cm x 58.4 cm (28.75 in. x 21 in. x 23 in.), Outer Dimensions (W x D x H): 79.4 cm x 59.7 cm x 79.1 cm (31.25 in. x 23.5 in.x 31.13 in.)
For Use With (Equipment)SH30717.02, SH30717.01. SH31047.01, SH31047.02
MaterialStainless Steel (304)
Product LineHyPerforma™, Smartainer™ II
Unit Size