Specifications
Capacity (Metric)1000 L
DimensionsInternal Dimensions (W x D x H) 114.3 cm x 91.4 cm x 114.8 cm (45 in. x 36 in. x 45.2 in.), Outer Dimensions (W x D x H) 120.7 cm x 97.8 cm x 135 cm (47.5 in. x 38.5 in. x 53.13 in.)
For Use With (Equipment)SH30969.05, SH30652.05
MaterialStainless Steel (304)
Product LineHyPerforma™, Smartainer™ II
Unit SizeEach