Specifications
Capacity (Metric)1500 L
DimensionsInternal Dimensions (W x D x H) 114.3 cm x 91.4 cm x 152.4 cm (45 in. x 36 in. x 60 in.), Outer Dimensions (W x D x H) 120.7 cm x 97.8 cmx 175.3 cm (47.5 in. x 38.5 in. x 69 in.)
For Use With (Equipment)SH30652.06, SH30969.06
MaterialStainless Steel (304)
Product LineHyPerforma™, Smartainer™ II
Unit SizeEach