Smart-Vue Pro Receivers
Smart-Vue Pro Receivers
Actual product may vary
Thermo Scientific™
Smart-Vue Pro Receivers
IT friendly LoRaWan provides easy set-up of a reliable, independent and secure wireless network
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionIncludes
SVPHWRLGW002Smart-Vue Pro Indoor Receiver for LoRa Modules (Enable Ethernet or WiFi Connectivity) 865-868 Mhz with Universal Power SupplyExternal Antenna, Universal Power Supply, LoRaWAN Flyer
SVPHWRLGW001Smart-Vue Pro Indoor Receiver for LoRa Modules (Enable Ethernet or WiFi Connectivity) 915 Mhz with Universal Power SupplyExternal Antenna, Universal Power Supply, LoRaWAN Flyer
2 Options
Catalog number SVPHWRLGW002
Price (USD)/ Each
-
Request A Quote
Description:
Smart-Vue Pro Indoor Receiver for LoRa Modules (Enable Ethernet or WiFi Connectivity) 865-868 Mhz with Universal Power Supply
Includes:
External Antenna, Universal Power Supply, LoRaWAN Flyer
LoRA is short for Long Range wireless Wide Area Network. It is a stand-alone dedicated network and increases connectivity range for IoT devices by 2 km in urban and 10 km in rural areas. This receiver pushes data to the Cloud by connecting to your local network via an Ethernet or Wi-Fi connection.
Long range wireless connectivity enables a single receiver to cover large areas, replacing the need for repeaters Dedicated wireless network requires only an internet connection, minimal IT support needed to set up. Support for up to 1000 data logging sensors per gateway
Specifications
TypeReceiver
Display TypeNo Display
CalibrationNot Calibrated
DescriptionSmart-Vue Pro Indoor Receiver for LoRa Modules (Enable Ethernet or WiFi Connectivity) 865-868 Mhz with Universal Power Supply
IncludesExternal Antenna, Universal Power Supply, LoRaWAN Flyer
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Range10 Miles Line of Sight, 2 to 3 Miles in Urban Environments
Unit SizeEach