Smart-Vue Pro Modules
Smart-Vue Pro Modules
Actual product may vary
Thermo Scientific™
Smart-Vue Pro Modules
Increase efficiency with multi-sensor wireless monitoring
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionIncludes
SVPHWRMOD014Two 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
SVPHWRMOD012Two 3.6 V Lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
Catalog number SVPHWRMOD014
Price (USD)
-
Request A Quote
Includes:
Two 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
Ask our AI about this Product
Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment
Bright 2.4 in. color touch screen with multiple views Alarms locally with real -time audio-visual alarms on the modules Audio visual alarms for local notification Communication locally via Bluetooth and to the system via LoRaLow AC power consumption through USB port access Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring Choice of wired and wireless sensors with automatic sensor detection User traceability with customized levels of user access with encrypted password protection
Specifications
TypeModule
Display TypeLCD
Battery Type3.6 V lithium battery
Battery Life2 years
IncludesTwo 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
related-product
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection