Smart-Vue Pro Alert Software
Smart-Vue Pro Alert Software
Actual product may vary
Thermo Scientific™
Smart-Vue Pro Alert Software
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SVPSWSA20100Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 100 PTS
SVPSWSA20005Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 5 PTS
SVPSWSA20020Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 20 PTS
SVPSWSA20050Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 50 PTS
SVPSWSA20250Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 250 PTS
SVPSWSA20500Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 500 PTS
SVPSWSA21000Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 1000 PTS
SVPSWSA21500Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 1500 PTS
SVPSWSA22000Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 2500 PTS
9 Options
Catalog number SVPSWSA20100
Price (USD)/ Each
-
Request A Quote
Description:
Smart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 100 PTS
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions, Smart-Vue™ Alert will automatically notify the user based notification preferences wherever you are (phone, text or email).
  • Add-on notification package through SMS and voice calls (Twillo)
  • 12 month license (post-activation)
  • License depends on the number of monitoring points desired
Specifications
TypeSmart-Vue Pro Software and Subscription Service
DescriptionSmart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 100 PTS
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
Unit SizeEach