Sensititre™ SWIN™ Software System
Sensititre™ SWIN™ Software System
Actual product may vary
Sensititre™ SWIN™ Software System
Sensititre™ SWIN™ Software System
Sensititre™ SWIN™ Software System
Sensititre™ SWIN™ Software System
Thermo Scientific™
Sensititre™ SWIN™ Software System
Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SW1202Sensititre SWIN Software Epidemiology Module
SW4000Sensititre SWIN Software, Complete
SW1100Labels, Barcode
SW130117 in. touch screen monitor
SW516Sensititre SWIN Bruker Interface
SW130222 in. touch screen monitor
Catalog number SW1202
Price (USD)
-
Request A Quote
Description:
Sensititre SWIN Software Epidemiology Module
Ask our AI about this Product
Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System. Simple touch screen navigation puts results at your fingertips while drop-down fields eliminate manual entry errors for all manual, semi-automated and fully automated reads.
Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Sensititre SWIN Software System.
  • Customizable Expert System provides three-tiered expert messages: Information, Modification (based on FDA, CLSI and EUCAST recommendations) and Warning Levels
  • Quality control module easily manages MIC results and broth information for inspection purposes
  • Automated alerts and report functionality enhance workflow and expedite release of results
Includes:
Microsoft Windows PC including dual hard drives, internal DVD burner, internal modem, barcode scanner, laser printer, remote access software, touchscreen monitor and SWIN Software.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionSensititre SWIN Software Epidemiology Module
Product TypeMicrobiology Media
Unit SizeEach
Intended use statement
The Sensititre SWIN software reads Sensititre (18-24 hr) susceptibility plates, generating a qualititative result of minimum inhibitory concentrations (MIC) and interpreting breakpoint (BP) results for non-fastidious and fastidious microorganisms. For use in combination with Sensititre instruments (Vizion and Aris) the results are used in a diagnostic workflow to aid clinicians in determining potential treatment options for patients suspected of having bacterial infections. The software is for professional use only and is not a companion diagnostic. Software generates an automatic result, but is interpreted manually.
Sensititre Pod Header
Explore our wide range of standard MIC plates and antimicrobials
Our latest Sensititre Plate Guides include everything you need to get to know the Sensititre System.
Download now ›
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Videos
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Share catalog number, name or link
      1x1 image pixel for data collection