Specifications
Chemical Name or MaterialEthanol
Melting Point-114°C
ColorColorless
AppearanceClear, Colorless Liquid
Boiling Point78°C
Quantity1 L
CAS7732-18-5
CAS64-17-5
PackagingSafe-Cote™ Amber Glass
Residue after Evaporation<=0.001%
Molecular FormulaC2 H6 O
WaterInclusive Between 2 - 6%
SynonymEthyl alcohol
Molecular Weight (g/mol)46.07
Percent Purity94 to 98%
GradeMolecular Biology
Unit SizeCase of 6