80% Ethanol, Molecular Biology Grade
This product ships via ground from US northeast.
80% Ethanol, Molecular Biology Grade
80% Ethanol, Molecular Biology Grade
80% Ethanol, Molecular Biology Grade
Thermo Scientific™

80% Ethanol, Molecular Biology Grade

Ready made ethanol/water blend prequalified for molecular biology research applications. | CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
Packaging:
Poly Bottle
Unit Size:
Each
Case of 4
Catalog number T08204K7
Price (USD)
220.00
Each
In stock
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Unit Size:
Each
Request bulk or custom format
Price (USD)
220.00
Each
Add to cart
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay of waterInclusive Between 18 - 22%
Protease activityPass Test
Substances Darkened by H2SO4Pass Test
DNasePass Test
RNasePass Test
View more
  • 80% ethanol and water blend
  • DNase-, RNase-, and protease-free
  • Ready made ethanol/water blend
  • Pre-qualified for molecular biology research applications
  • Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
  • Tested for use with Applied Biosystems™ MagMAX™ kits
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection