Informational:NEW - Major Software Update Available! — Cell Cycle Analysis can be performed using the Tali® Image-Based Cytometer
Invitrogen™

Tali™ Calibration Beads

Catalog number: T10790
Have Questions?
Invitrogen™

Tali™ Calibration Beads

Catalog number: T10790
Have Questions?
Catalog Number
T10790
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T107901 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This product contains three tubes of beads which are used to properly calibrate and align the Tali™ Image-Based Cytometer.

Contents:

• Tali™ Alignment Beads,
• Tali™ Green Calibration Beads
• Tali™ Red Calibration Beads

The calibration function of the Tali™ Image-Based Cytometer recalculates the dynamic range and checks the alignment of the camera. Proper alignment and calibration of the Tali™ Image-Based Cytometer ensures optimal performance of the instrument. It is recommended that alignment and calibration be performed every three months.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Calibration Bead
For Use With (Equipment)
Tali™ Image-Based Cytometer
Color
Red, Green
Shipping Condition
Room Temperature
Format
Kit
Product Line
Tali™

Contents & Storage

Tali™ - Green Calibration Beads: Store 2 – 6°C
Tali™ - Red Calibration Beads: Store 2 – 6°C
Tali™ - Alignment Beads; Store 2 – 6°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text