You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
ChromaCare™ LC-MS Instrument Flush Solution, Thermo Scientific™
Element Search
Structure Search
This product ships via ground from US northeast.
Thermo Scientific™
ChromaCare™ LC-MS Instrument Flush Solution, Thermo Scientific™
ChromaCare™ LC-MS Instrument Flush Solution is designed to prepare LC/MS instruments for start-up. By reducing background noise, this solution facilitates instrument installation and preventative maintenance routines.
Have Questions?
Change view
Quantity:
1 L
Packaging:
Borosilicate glass bottle
Unit Size:
Each
Case of 6
Try the chem dex
TM
web app
Catalog number
T111101000
Price (USD)
/ Each
111.65
Online exclusive
124.00
Save 12.35 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
Borosilicate glass bottle
Unit Size:
Each
Request bulk or custom format
Price (USD)
/ Each
111.65
Online exclusive
124.00
Save 12.35 (10%)
Add to cart
ChromaCare™ LC-MS Instrument Flush Solution, Thermo Scientific™
Catalog number
T111101000
Price (USD)
/ Each
111.65
Online exclusive
124.00
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS
75-05-8; 67-56-1; 67-63-0; 7732-18-5
Specifications
Description
LC-MS Instrument Flush Solution
CAS
75-05-8; 67-56-1; 67-63-0; 7732-18-5
Packaging
Borosilicate glass bottle
Quantity
1 L
Color
Colorless
View more
Designed by Thermo Scientific to maximize cleaning while preserving instrument integrity in Vanquish™ and UltiMate™ UHPLC systems with QQQ MS
Comprised of a unique blend of polar protic and aprotic solvents for removing a wide variety of contaminants
Particularly suitable for new LC/MS and UHPLC/MS installations to rapidly stabilize baseline
A blend of 25% acetonitrile, 25% methanol, 25%, water, and 25% 2-propanol (IPA)
Offered in 1L borosilicate bottles and 2.5L amber glass bottles
RUO – Research Use Only