TE Buffer (20X), RNase-free
TE Buffer (20X), RNase-free
TE Buffer (20X), RNase-free
TE Buffer (20X), RNase-free
Invitrogen™

TE Buffer (20X), RNase-free

Essential to success of RiboGreen™ RNA quantitation assays
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Catalog NumberQuantity
T11493Promo Image100 mL
Catalog number T11493
Price (USD)
140.65
Online Exclusive
147.00
Save 6.35 (4%)
100 ml
In stock
Add to cart
Quantity:
100 mL
Price (USD)
140.65
Online Exclusive
147.00
Save 6.35 (4%)
100 ml
Add to cart
RNase-free TE Buffer (20X) is an essential component of Qubit and Quant-iT assays, including PicoGreen, RiboGreen, and Qubit dsDNA HS. It is offered here as a standalone solution to support quantification of dsDNA, ssDNA and RNA.

Note: For dilution instructions, storage considerations, and assay compatibility information please see the Product Information Sheet (Pub. No. MAN1000402) below.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20X
FormLiquid
Product TypeTE Buffer
Quantity100 mL
Shipping ConditionRoom Temperature
Unit Size100 ml
Contents & Storage
Store at room temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link