Informational:Get the sharpest, brightest images. Try new Invitrogen™ SlowFade™ Glass antifade RIMS mountant with refractive index of 1.52. Learn more »
Invitrogen™

Texas Red™ 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt (Texas Red™ DHPE)

Catalog number: T1395MP
Have Questions?
Invitrogen™

Texas Red™ 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt (Texas Red™ DHPE)

Catalog number: T1395MP
Have Questions?
Catalog Number
T1395MP
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T1395MP1 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The phospholipid, Texas Red™ 1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt (Texas Red™ DHPE) is labeled on the head group with the bright, red-fluorescent Texas Red™ dye (excitation/emission maxima ∼595/615 nm).

To make a concentrated stock solution from 1–2 mg/mL, dissolve in 100% ethanol. Sonication may be necessary to aid dispersion. Store frozen, desiccated and protected from light. The gradual accumulation of moisture from the atmosphere may promote aggregation of the reagent over time.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name or Material
Phospholipids
Recommended Storage
Store in freezer (-5°C to -30°C) and protect from light.
Quantity
1 mg
Molecular Weight (g/mol)
1381.85
Physical Form
Solid
Product Line
Texas Red
Excitation/Emission
583/601 nm
Unit Size
1 mg

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text