FM™ 1-43 Dye (N-(3-Triethylammoniumpropyl)-4-(4-(Dibutylamino) Styryl) Pyridinium Dibromide)
FM&trade; 1-43 Dye (<i>N</i>-(3-Triethylammoniumpropyl)-4-(4-(Dibutylamino) Styryl) Pyridinium Dibromide)
Invitrogen™

FM™ 1-43 Dye (N-(3-Triethylammoniumpropyl)-4-(4-(Dibutylamino) Styryl) Pyridinium Dibromide)

FM 1-43 membrane probe is an excellent reagent both for identifying actively firing neurons and for investigating the mechanisms ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
T31631mg
T3535610 x 100μg
Catalog number T3163
Price (USD)/ 1 mg
480.65
Online exclusive
495.00 
Save 14.35 (3%)
-
Add to cart
Quantity:
1mg
Price (USD)/ 1 mg
480.65
Online exclusive
495.00 
Save 14.35 (3%)
Add to cart
FM™ 1-43 Dye (N-(3-Triethylammoniumpropyl)-4-(4-(Dibutylamino) Styryl) Pyridinium Dibromide)
Catalog numberT3163
Price (USD)/ 1 mg
480.65
Online exclusive
495.00 
Save 14.35 (3%)
-
Add to cart
FM 1-43 membrane probe is an excellent reagent both for identifying actively firing neurons and for investigating the mechanisms of activity-dependent vesicle cycling. This water-soluble dye, which is nontoxic to cells and virtually nonfluorescent in aqueous medium, is believed to insert into the outer leaflet of the cell membrane where it becomes intensely fluorescent. In a neuron that is actively releasing neurotransmitters, the dye becomes internalized within the recycled synaptic vesicles and the nerve terminals become brightly stained. The nonspecific staining of cell-surface membranes can simply be washed off prior to viewing. FM 1-43 membrane probe is also available as 1 mg in a single vial (T-3163). FM 1-43FX membrane probe (F-35355), an analog of FM 1-43 membrane probe that can be fixed in place using aldehyde-based fixatives, is also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange
For Use With (Equipment)Fluorescence Microscope
Quantity1mg
Detection MethodFluorescence
Product LineFM™
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeDye
SubCellular LocalizationPlasma Membrane
Unit Size1 mg
Contents & Storage
Store at room temperature and protect from light.