Invitrogen™

FM™ 4-64 Dye (N-(3-Triethylammoniumpropyl)-4-(6-(4-(Diethylamino) Phenyl) Hexatrienyl) Pyridinium Dibromide)

Catalog number: T3166
Have Questions?
Invitrogen™

FM™ 4-64 Dye (N-(3-Triethylammoniumpropyl)-4-(6-(4-(Diethylamino) Phenyl) Hexatrienyl) Pyridinium Dibromide)

Catalog number: T3166
Have Questions?
Catalog Number
T3166
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 100 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T31661 mg
-
T1332010 x 100 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The styryl dye FM 4-64 has been reported to selectively stain yeast vacuolar membranes with red fluorescence (excitation/emission maxima ∼515/640 nm). This lipophilic dye is an important tool for visualizing vacuolar organelle morphology and dynamics, for studying the endocytic pathway and for screening and characterizing yeast endocytosis mutants. FM 4-64 is also available in special packaging with 10 vials of 100 μg each (T-13320).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Red
For Use With (Equipment)
Fluorescence Microscope
Quantity
1mg
Detection Method
Fluorescent
Product Line
FM™
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Dye
SubCellular Localization
Plasma Membrane

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text