Invitrogen™

TC-ReAsH™ II In-Cell Tetracysteine Tag Detection Kit (Red Fluorescence), for live-cell imaging

Catalog number: T34562
Have Questions?
Invitrogen™

TC-ReAsH™ II In-Cell Tetracysteine Tag Detection Kit (Red Fluorescence), for live-cell imaging

Catalog number: T34562
Have Questions?
Catalog Number
T34562
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T345621 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
TC-ReAsH™ II In-Cell Tetracysteine Tag Detection Kit contains an expression tag-based fluorescence labeling reagent for live-cell labeling. Mammalian cell lines expressing a protein fused to a tetracysteine tag (CCPGCC) can be labeled with the red-fluorescent ReAsH-EDT2 reagent contained in the kit. The tagged protein is fluorescent only when the labeling reagent is added. BAL wash buffer replaces the previously supplied Disperse Blue 3 and EDT-based wash buffer as an olfactorily more agreeable reagent that yields superior signal to noise.


The kit contains ReAsH-EDT2 labeling reagent (store at -20°C, protected from light) and BAL wash buffer (stored at 4°C). When stored as directed, the kit is stable for 6 months.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Tetracysteine Tag Detection Kit
Detection Method
Fluorescent
For Use With (Equipment)
Fluorescence Microscope
Color
Red
Label or Dye
ReAsH
Shipping Condition
Wet Ice
Format
Kit
Product Line
TC-ReAsH II

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text