The thiol-reactive Texas Red™ C2 maleimide can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity
5 mg
Product Type
Maleimide
Chemical Reactivity
Thiol
Shipping Condition
Room Temperature
Reactive Moiety
Maleimide
Label or Dye
Texas Red™
Label Type
Classic Dyes
Product Line
Texas Red™
Emission
615
Excitation
595
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.