The primary aliphatic amine of the red-fluorescent Texas Red™ hydrazide can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity
5 mg
Product Type
Hydrazide
Chemical Reactivity
Carboxylic Acid, Ketone, Aldehyde
Shipping Condition
Room Temperature
Reactive Moiety
Amine, Hydrazide
Label or Dye
Texas Red™
Label Type
Classic Dyes
Product Line
Texas Red™
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.