Tetramethylrhodamine, methyl ester (TMRM) is a cell-permeant, cationic, red-orange fluorescent dye that is readily sequestered by active mitochondria.Visualize staining yourRead more
Have Questions?
Catalog Number
Quantity
T668
25 mg
Catalog number T668
Price (USD)
271.65
Online Exclusive
280.00
Save 8.35 (3%)
25 mg
Add to cart
Quantity:
25 mg
Price (USD)
271.65
Online Exclusive
280.00
Save 8.35 (3%)
25 mg
Add to cart
Tetramethylrhodamine, methyl ester (TMRM) is a cell-permeant, cationic, red-orange fluorescent dye that is readily sequestered by active mitochondria.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Quantity25 mg
Shipping ConditionRoom Temperature
Sub Cellular LocalizationMitochondria
ColorRed-Orange
For Use With (Equipment)Fluorescence Microscope
Product TypeTMRM
Unit Size25 mg
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Frequently asked questions (FAQs)
I am seeing high background outside of my neuronal cells when using membrane potential indicators. What can I do to reduce background?
If you use our FluoVolt Membrane Potential Kit (Cat. No. F10488), the kit provides a background suppressor to reduce this problem. For other indicators, consider the use of BackDrop Background Suppressor (Cat no. R37603, B10511, and B10512).
What is the difference between fast and slow-response membrane potential probes?
Molecules that change their structure in response to the surrounding electric field can function as fast-response probes for the detection of transient (millisecond) potential changes. Slow-response dyes function by entering depolarized cells and binding to proteins or membranes. Increased depolarization results in additional dye influx and an increase in fluorescence, while hyperpolarization is indicated by a decrease in fluorescence. Fast-response probes are commonly used to image electrical activity from intact heart tissues or measure membrane potential changes in response to pharmacological stimuli. Slow-responding probes are often used to explore mitochondrial function and cell viability.
What type of membrane potential indicators do you offer and how should I choose one for my experiment?
A membrane potential indicator selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-viability-and-regulation/ion-indicators/membrane-potential-indicators.html).
I am testing mitochondrial membrane potential, but my untreated cells are fluorescing, and I'm not seeing a significant difference in my test sample.
Regardless of which dye you use - tetramethylrhodamine, methyl ester (TMRM), JC-1 or MitoTracker - untreated cells will fluoresce. It's just that cells with reduced mitochondrial membrane potential will fluoresce less. It is the degree of change which is important. JC-1 dye not only changes intensity, but has a ratiometric spectral change in excitation and emission. It is very important to have an untreated control as well as a positive control treated with a mitochondrial membrane potential destabilizer, such as CCCP or FCCP. Most mitochondrial stains are only for use with live cells, as the signal will not be retained to the same degree with fixation.
Spatially organised mitochondrial calcium uptake through a novel pathway in chick neurones.
Authors:Coatesworth W, Bolsover S
Journal:Cell Calcium
PubMed ID:16338004
'A brief depolarisation of chick sensory neurones evokes a calcium increase in mitochondria that peaks 1-2s after the depolarisation event and then decays over tens of seconds. Peripheral mitochondria take up more calcium than do central ones, even when the cytosolic calcium increase is spatially homogeneous. The calcium influx into ... More
Tissue transglutaminase is a multifunctional BH3-only protein.
Authors:Rodolfo C, Mormone E, Matarrese P, Ciccosanti F, Farrace MG, Garofano E, Piredda L, Fimia GM, Malorni W, Piacentini M
Journal:J Biol Chem
PubMed ID:15485857
'Tissue transglutaminase (TG2) protein accumulates to high levels in cells during early stages of apoptosis both in vivo and in vitro. The analysis of the TG2 primary sequence showed the presence of an eight amino acid domain, sharing 70% identity with the Bcl-2 family BH3 domain. Cell-permeable peptides, mimicking the ... More
Methods for assessing autophagy and autophagic cell death.
Authors:Tasdemir E, Galluzzi L, Maiuri MC, Criollo A, Vitale I, Hangen E, Modjtahedi N, Kroemer G,
Journal:Methods Mol Biol
PubMed ID:18425442
'Autophagic (or type 2) cell death is characterized by the massive accumulation of autophagic vacuoles (autophagosomes) in the cytoplasm of cells that lack signs of apoptosis (type 1 cell death). Here we detail and critically assess a series of methods to promote and inhibit autophagy via pharmacological and genetic manipulations. ... More
Trafficking of ganglioside GD3 to mitochondria by tumor necrosis factor-alpha.
Authors:García-Ruiz C, Colell A, Morales A, Calvo M, Enrich C, Fernández-Checa JC
Journal:J Biol Chem
PubMed ID:12118012
'The interaction of mitochondria with proapoptotic proteins activates apoptosis pathways. Previous findings have identified ganglioside GD3 (GD3) as an emerging apoptotic lipid intermediate that targets mitochondria in response to death signals. Using immunoelectron and laser scanning confocal microscopy, we characterize the trafficking of GD3 to mitochondria in response to tumor ... More
Unreliability of the cytochrome c-enhanced green fluorescent fusion protein as a marker of cytochrome c release in cells that overexpress Bcl-2.
Authors:Unkila M, McColl KS, Thomenius MJ, Heiskanen K, Distelhorst CW
Journal:J Biol Chem
PubMed ID:11489892
'A cytochrome c-enhanced green fluorescent protein chimera (cyt-c.EGFP) was used to monitor the release of cytochrome c from mitochondria in Bcl-2-negative and Bcl-2-positive MDA-MB-468 breast cancer cells. A comparison was made with the intracellular distribution of endogenous cytochrome c based on Western blotting of cell fractions and immunocytochemistry. The release ... More