Invitrogen™

TetraSpeck™ Microspheres, 4.0 μm, fluorescent blue/green/orange/dark red

Catalog number: T7283
Have Questions?
Invitrogen™

TetraSpeck™ Microspheres, 4.0 μm, fluorescent blue/green/orange/dark red

Catalog number: T7283
Have Questions?
Catalog Number
T7283
also known as T-7283
Unit Size
0.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T7283
also known as T-7283
0.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The 4.0 μm TetraSpeck™ microspheres are stained throughout with four different fluorescent dyes, yielding beads that each display four well-separated excitation/emission peaks - 360/430 nm (blue), 505/515 nm (green), 560/580 nm (orange) and 660/680 nm (dark red). These microspheres can greatly facilitate the calibration of conventional fluorescence microscopes, confocal laser scanning microscopes, and associated image-processing equipment for both scientific and commercial imaging, especially for multicolor applications.

See our full collection of microscope calibration reagents ›

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibration Type
Confocal Microscope Calibration, Fluorescence Microscope Calibration
Color
Orange, Dark Red, Blue, Green
Product Type
Microsphere
Format
Suspension Beads
Product Line
TetraSpeck
Quantity
0.5 mL
Shipping Condition
Room Temperature
Diameter (Metric)
4 μm
Unit Size
0.5 mL

Contents & Storage

Store in refrigerator 2°C to 8°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text