Invitrogen™

Texas Red™-X Phalloidin

Catalog number: T7471
Have Questions?
Invitrogen™

Texas Red™-X Phalloidin

Catalog number: T7471
Have Questions?
Catalog Number
T7471
Unit Size
300 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T7471300 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Texas Red-X phalloidin is a high-affinity F-actin probe conjugated to our bright, photostable, red fluorescent Texas Red-X dye.

Selectively stains F-actin
Outstanding fluorescence performance
Excitation/Emission: 591/608 nm
Superior to antibody staining
Optimal for fixed and permeabilized samples

Get Superior Results in Actin Staining Studies
Phalloidin is a bicyclic peptide belonging to a family of toxins isolated from the deadly Amanita phalloides 'death cap' mushroom and is commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has virtually identical binding properties with actin from different species including plants and animals. Phalloidin binds F-actin with high selectivity while Texas Red-X provides red fluorescence with superior brightness. Demonstrating very little nonspecific staining, Texas Red-X phalloidin allows high-contrast discrimination of actin staining.

Use in Multiple Applications
Texas Red-X phalloidin can be used to visualize and quantitate F-actin in tissue sections, cell cultures, or cell-free preparations. Texas Red-X phalloidin staining is fully compatible with other fluorescent stains used in cellular analyses including fluorescent proteins, Qdot nanocrystals, and Alexa Fluor conjugates including secondary antibodies.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links
Read about labeling F-Actin with phallotoxins.
Create a Cellular Masterpiece with Alexa Fluor Phalloidin using the Cell Paint Tool.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation Wavelength Range
591⁄608
Dye Type
Texas Red™
Product Line
Texas Red™
Shipping Condition
Room Temperature
Label Type
Classic Dyes
Product Type
Phalloidin
SubCellular Localization
Actin, Cytoskeleton

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text