Invitrogen™

FM™ 2-10 (N-(3-Triethylammoniumpropyl)-4-(4-(Diethylamino)styryl)Pyridinium Dibromide)

Catalog number: T7508
Have Questions?
Invitrogen™

FM™ 2-10 (N-(3-Triethylammoniumpropyl)-4-(4-(Diethylamino)styryl)Pyridinium Dibromide)

Catalog number: T7508
Have Questions?
Catalog Number
T7508
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
T75085 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The FM™ 2-10 membrane probe is an excellent reagent both for identifying actively firing neurons and for investigating the mechanisms of activity-dependent vesicle cycling. This water-soluble dye, which is nontoxic to cells and virtually nonfluorescent in aqueous medium, is believed to insert into the outer leaflet of the cell membrane where it becomes intensely fluorescent. In a neuron that is actively releasing neurotransmitters, the dye becomes internalized within the recycled synaptic vesicles and the nerve terminals become brightly stained.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Orange
For Use With (Equipment)
Fluorescence Microscope
Detection Method
Fluorescent
Product Line
FM™
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Membrane Stain
SubCellular Localization
Plasma Membrane

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text