Thermo Scientific™

TruBio™ Discovery Automation System Bundle, 3 L HyPerforma Glass Bioreactor, TrupH and TruDO sensors

Número de catálogo: TBD1-HPGLS3-TRU
Thermo Scientific™

TruBio™ Discovery Automation System Bundle, 3 L HyPerforma Glass Bioreactor, TrupH and TruDO sensors

Número de catálogo: TBD1-HPGLS3-TRU
Número de catálogo
TBD1-HPGLS3-TRU
Tamaño de la unidad
Each
Precio (MXN)
Número de catálogoTamaño de la unidadPrecio (MXN)
TBD1-HPGLS3-TRUEachContáctenos ›
Descripción general del producto
Figuras
Videos
Recomendaciones
Recomendaciones
Documentos
Preguntas frecuentes
Citas y referencias
Información adicional
Recomendaciones
The Thermo Scientific TruBio Discovery Automation System Bundle with 3 L HyPerforma Glass Bioreactor, HyPerforma G3Lab Controller, and TruBio Bioprocess Control Software powered by DeltaV Discovery Distributed Control Platform by Emerson is an out-of-the-box, workstation-based, scalable automation solution for research and process development applications.

Features of the TruBio Discovery Automation System:
Transferability of software—from research and development, clinical, and commercial production processes
Scalability—standardizing processes on TruBio Discovery system early in process development sets up your lab for easy data transfer during scale-up and commercialization
An open architecture solution—can be packaged with Thermo Scientific bench-scale equipment or third-party vessels

Simplifying tech transfer from research and process development into clinical trials and commercialization, this automation package can be scaled up to operate up to eight different bioreactors simultaneously. Based on an open architecture approach, these bioreactor systems can be identical or differ by supplier, unit size, etc., allowing for greater efficiency in your processes with a smaller footprint. Unlike systems based on programmable logic control (PLC), the TruBio Discovery system is based on a distributed control system (DCS), so data and recipes can be stored and easily transferred as you scale-up your processes. Users are able to retain valuable historical data, recipes, and batch records.

This automation system bundle includes:
• Automation software package for one vessel
• 3 L HyPerforma Glass Bioreactor
• HyPerforma G3Lab Controller
• Mass flow controller (MFC) for one vessel
• Adapter kit for one vessel
• Sensor kit

Applications
• Cell culture
• Process development-scale cell expansion
• Early-phase clinical drug development

For expansion and further configuration of this automation system bundle, please request a quote and speak to your sales representative. This bundle has the capability to expand to support eight vessels of any configuration.

Especificaciones

Material
Glass (Bioreactor)
Línea de productos
TruBio™
Product Type Specs
Discovery Automation System Bundle

Contenido y almacenamiento

• Automation software bundle for one vessel
• 3 L HyPerforma Glass Bioreactor
• G3Lab Controller
• Mass Flow Controller (MFC) for one vessel
• Adapter kit for one vessel
• Sensor kit

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text