Search Thermo Fisher Scientific
Thermo Scientific™ Tensile Sample Holder for Thermo Scientific™ Phenom XL. Tensile testing is one of the simplest and most widely used mechanical tests. By measuring the force required to elongate a specimen to breaking point, material properties can be determined that will allow designers and quality managers to predict how materials and products will behave in their intended applications.
Unit Size | Each |
Número do catálogo | Especificações | Tamanho da unidade | Preço (BRL) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TENSILESAMPLEHOLD | Each | Requisite uma Cotação | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Applications for the Tensile Sample Holder:
Industrial applications are manifold and located in aerospace and automotive industry, packaging/paper industry or even in the area of pharmaceuticals.