Sterilin™ Pipette Bulbs
Sterilin™ Pipette Bulbs
Thermo Scientific™
Sterilin™ Pipette Bulbs
Transfer liquids safely using glass pipettes and these natural rubber pipette bulbs.
Have Questions?
Catalog NumberDescription
TET0102Pipette bulbs; No. 10 Teat (Case of 100)
Catalog number TET0102
Price (EUR)
-
Request A Quote
Description:
Pipette bulbs; No. 10 Teat (Case of 100)
Ask our AI about this Product
Transfer liquids safely using glass pipettes and these natural rubber Sterilin™ Pipette Bulbs.
  • Ideal for use with glass Pasteur pipettes
  • Manufactured from natural rubber
  • Volume drawn up: 0.22 cm3
Ordering Information
  • Not available in all geographic regions
Specifications
DescriptionPipette bulbs; No. 10 Teat (Case of 100)
For Use With (Equipment)Ideal for use with glass Pasteur pipettes
For Use WithGlass Pasteur Pipette
MaterialNatural Rubber
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
M494600Certificate of ComplianceDec 03, 2024TET0102
M548630Certificate of ComplianceDec 03, 2024TET0102
M475800Certificate of ComplianceDec 03, 2024TET0102
M479310Certificate of ComplianceDec 03, 2024TET0102
M484000Certificate of ComplianceDec 03, 2024TET0102
5 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection