Search Thermo Fisher Scientific
Leverage the reliable and robust technology of the Thermo Scientific™ TRACE™ 1310 Gas Chromatograph to analyze transformer insulating oils for dissolved gases.
Catalog Number | Description |
---|---|
TOGAMAC010022 also known as TMP00148 | Transformer oil gas analyzer with sulfur bypass |
TOGAMAC010011 | Transformer oil gas analyzer |
Automate the analysis of dissolved gases in transformer insulating oils using the Thermo Scientific™ TRACE™ GC for Dissoved Gases. This gas chromatography system is based on the robust and reliable Thermo Scientific™ TRACE™ 1310 series gas chromatograph. It includes all of the components required to measure gas samples from transformer oil and is fully compliant with ASTM D3612 Method A or C. The system is delivered pre-configured and tested to enable fast startup and commissioning. Documentation includes startup procedures, troubleshooting guide, instrument method, and recommended spare parts.
Ready to install and run samples