TruDefender™ FTX Handheld FTIR for Chemical Identification
TruDefender™ FTX Handheld FTIR for Chemical Identification
Thermo Scientific™

TruDefender™ FTX Handheld FTIR for Chemical Identification

Mitigate the potential risk of an unknown chemical threat with fast, accurate identification of explosives or hazardous materials.
Have Questions?
Catalog NumberDescription
TRUDEFENDERFTXTruDefender FTX Handheld FTIR for Chemical Identification
Catalog number TRUDEFENDERFTX
Price (USD)
-
Request A Quote
Description:
TruDefender FTX Handheld FTIR for Chemical Identification

When faced with an unknown chemical threat, every second matters. Fast, accurate identification of potential explosives or hazardous materials is key to ensuring the safety of hazmat teams and other first responders, as well as the community.

Thermo Scientific™ TruDefender™ FTX handheld chemical identification analyzers enable effective incident response, providing fast, actionable information to mitigate potential threats. Based on FTIR spectroscopy, TruDefender FTX provides easy sampling of solid and liquid chemicals in the field using the self-contained anvil sampling head.

The TruDefender FTXi analyzer is not available in all countries. Contact us for availability. Fast, Accurate Identification Returns results in seconds, even for complex mixtures. Improved Sampling Capability New anvil sampling head for easier sampling and decontamination. Anvil rotates for easy sample placement and cleaning. Variable Pressure Knob Allows technician to determine appropriate pressure for a given sample. Easy to Clean Highly polished, rounded corners. No crevasses or friction points to trap debris or explosives residue. Liquid Containment Moat Helps prevent sample spills. Flexible Sampling The reversible sampling accessory provides environmental protection for volatile liquids and enables easy sampling of prills. Easy to Use Intuitive, menu-driven interface for fast training and proficiency.
Specifications
Product TypeHandheld FTIR Spectrometer
Battery TypeRemovable and rechargeable lithium ion battery or 123a (e.g. SureFire) batteries; > 4 hours operation
Certifications/ComplianceIndependently tested for MIL-STD-810G and IP67 certification
DescriptionTruDefender FTX Handheld FTIR for Chemical Identification
Dimensions (L x W x H)8.9 x 4.5 x 2.1 in. (22.61 x 11.42 x 5.33 cm)
Height (English)2.1 in.
Height (Metric)5.33 cm
IP RatingIP67
LanguagesEnglish, Chinese, German, Russian (Optional)
Length (English)8.9 in.
Length (Metric)22.61 cm
Optical DesignSolid Diamond Crystal ATR
Spectral Range4,000 cm-1 to 650 cm-1
Temperature Range (English)-4 °F to +104°F
Temperature Range (Metric)-20°C to +40°C
Weight (English)3.12 lb.
Weight (Metric)1.41 kg
Width (English)4.5 in.
Width (Metric)11.42 cm
Unit SizeEach

Share catalog number, name or link