Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Currently Unavailable
Eliminate dangerous counterfeit or substandard products from your supply chain. The Thermo Scientific™ TruScan™ GP Analyzer provides quick, easy to use raw material identification and finished product inspection to screen out counterfeit substances and reduce supply chain risk. Building on the proven capability of our flagship Thermo Scientific™ TruScan™ RM Analyzer, the TruScan GP Analyzer is designed to provide cost-effective material quality control to pharmaceutical manufacturers and chemical companies in emerging global markets.
Barcode Supported Symbologies | Most linear and 2D standards |
Battery Type | Rechargeable internal lithium ion battery > 3 hours operation |
Certifications/Compliance | FDA 1040, 21 CFR part 11, CE certification |
Collection Optics | NA = 0.33, 18 mm working distance; 0.2 to 2.5 mm spot size |
Description | TruScan GP Material Identification Analyzer |
Dimensions (L x W x H) | 8.2 x 4.2 x 1.7 in. (20.8 x 10.7 x 4.3 cm) |
Exposure | Automatic modes (12 ms minimum) |
Height (English) | 1.7 in. |
Height (Metric) | 4.3 cm |
Laser Excitation Wavelength | 785 nm ± 0.5 nm, 2 cm-1 line width, stability <0.1 cm-1 |
Laser Output | 250 mW ±25 |
Length (English) | 8.2 in. |
Length (Metric) | 20.8 cm |
Optional Accessories | Vial and tablet holders |
Power Supply | DC wall adapter 100/240 V, 50/60 Hz |
Spectral Range | 250 to 2875 cm-1 |
Spectral Resolution | 8 to 10.5 cm-1(FWHM) across range |
Temperature Range (Metric) | -20°C to 40°C |
Weight (English) | 2 lb. |
Weight (Metric) | 0.9 kg |
Width (English) | 4.2 in. |
Width (Metric) | 10.7 cm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TRUSCANGP | Each | TruScan GP Material Identification Analyzer | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|