PTFE/Rubber Laminated Discs
PTFE/Rubber Laminated Discs
Actual product may vary
Thermo Scientific™
PTFE/Rubber Laminated Discs
For a highly inert and unreactive seal use Thermo Scientific™ PTFE/Rubber Laminated Discs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure Size
TS-1241212 mm
TS-1241818 mm
TS-1242222 mm
3 Options
Catalog number TS-12412
Price (USD)/ Pack of 72
80.65
Add to cart
Closure Size:
12 mm
Price (USD)/ Pack of 72
80.65
Add to cart
PTFE/Rubber Laminated Discs
Catalog numberTS-12412
Price (USD)/ Pack of 72
80.65
-
Add to cart
For a highly inert and unreactive seal use Thermo Scientific™ PTFE/Rubber Laminated Discs. Constructed of white pharmaceutical rubber with 5 mils of PTFE coating bonded to one side, they are excellent for use as cap liners when a highly inert nonreactive seal is desired. Discs are autoclavable with no loss of integrity after heating above 100 °C for five hours. Several sizes are available to fit Thermo Scientific™ Reacti-Vial™ Small Reaction Vials and Thermo Scientific™ Screw Cap Septum Vials.
Additional Product Information
  • Disc thickness is approximately 60 mils
  • Discs are rigid and more difficult to puncture than Thermo Scientific™ PTFE/Silicone Discs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description12mm
MaterialPTFE, Rubber
AutoclavableAutoclavable
Quantity72 Pk.
Thickness (Metric)60 mil
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeDisc
Closure Size12 mm
For Use With (Equipment)Small Reaction Vial
Unit SizePack of 72