Reacti-Vial™ Small Glass Reaction Vials
Reacti-Vial™ Small Glass Reaction Vials
Thermo Scientific™

Reacti-Vial™ Small Glass Reaction Vials

Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric)QuantityIncludes
TS-13097Amber1 mL12 Pk.Open-Top Screw Caps and PTFE/Rubber Discs
TS-13099Amber5 mL12 Pk.Open-Top Screw Caps and PTFE/Rubber Discs
TS-13100Clear0.1 mL12 Pk.Open-Top Screw Caps and PTFE/Rubber Discs
TS-13220Clear0.3 mL12 Pk.Open-Top Screw Caps and PTFE/Rubber Discs
TS-13221Clear1 mL12 Pk.Open-Top Screw Caps and PTFE/Rubber Discs
TS-13222Clear3 mL12 Pk.Open-Top Screw Caps and PTFE/Rubber Discs
TS-13225Clear10 mL12 Pk.Open-Top Screw Caps and PTFE/Rubber Discs
Catalog number TS-13097
Price (USD)/ Each
-
Request A Quote
Color:
Amber
Volume (Metric):
1 mL
Quantity:
12 Pk.
Includes:
Open-Top Screw Caps and PTFE/Rubber Discs

Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module. An internal cone makes small sample handling in these vials easy and convenient. The extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions. Reacti-Vials are available in a variety of sizes of clear and amber glass, and are supplied with Thermo Scientific™ Open-Top Screw Caps and PTFE/Rubber Discs.

Applications
  • Residue isolation
  • Derivative preparation
  • Maximum sample retrieval
  • Moisture protection
  • Sample storage
  • Precipitations
  • Centrifugations
  • Solvent evaporation

Additional Product Information

  • Supplied 12/pack, complete with Thermo Scientific Open-Top Screw Caps and Thermo Scientific PTFE/Rubber Laminated Discs
  • Available in several sizes
  • Thermo Scientific PTFE/Silicone Discs ordered separately
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
ColorAmber
MaterialGlass
Volume (Metric)1 mL
IncludesOpen-Top Screw Caps and PTFE/Rubber Discs
Thread StyleInternal
Product LineReacti-Vial
Volume (Metric) Total1 mL
Closure TypeScrew Cap
Quantity12 Pk.
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeVial
Closure Size13-425
Unit SizeEach