Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.
For Use With | GC |
Standard Concentration | 98 to 100, 0.1 to 1% |
Item Description | BSTFA +1% TMCS; 10 x 1 mL ampules |
Storage | 2°C to 8°C |
Quantity | 10 x 1 mL ampules |
Standard Components | BSTFA +1% TMCS |
Unit Size | Each |
For Use With | GC |
Standard Concentration | 98 to 100, 0.1 to 1% |
Item Description | BSTFA +1% TMCS; 10 g |
Storage | 2°C to 8°C |
Quantity | 10 g |
Standard Components | BSTFA +1% TMCS |
Unit Size | Each |
For Use With | GC |
Standard Concentration | 98 to 100, 0.1 to 1% |
Item Description | BSTFA +1% TMCS; 25 g |
Storage | 2°C to 8°C |
Quantity | 25 g |
Standard Components | BSTFA +1% TMCS |
Unit Size | Each |
For Use With | GC |
Standard Concentration | 98 to 100, 0.1 to 1% |
Item Description | BSTFA +1% TMCS; 100 g |
Storage | 2°C to 8°C |
Quantity | 100 g |
Standard Components | BSTFA +1% TMCS |
Unit Size | Each |
Item Description | BSTFA +10% TMCS; 10 x 1 mL Ampules |
Storage | 2°C to 8°C |
Quantity | 10 x 1 mL Ampules |
Standard Components | BSTFA +10% TMCS |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Standard Components | Quantity | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TS-38831 | Each | BSTFA +1% TMCS | 10 x 1 mL ampules | 177.38 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TS-38832 | Each | BSTFA +1% TMCS | 10 g | 131.71 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TS-38833 | Each | BSTFA +1% TMCS | 25 g | 168.24 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TS-38834 | Each | BSTFA +1% TMCS | 100 g | 410.85 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TS-38840 | Each | BSTFA +10% TMCS | 10 x 1 mL Ampules | 333.91 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|