BSA Silylation Reagent
BSA Silylation Reagent
Actual product may vary
Thermo Scientific™
BSA Silylation Reagent
Produce volatile, stable TMS derivatives quickly and quantitatively under mild conditions with Thermo Scientific™ BSA Silylation Reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TS-38839100 g
TS-3883825 g
2 Options
Catalog number TS-38839
Price (USD)/ Each
-
Request A Quote
Quantity:
100 g
Derivatize analytes quickly and quantitatively under mild conditions using Thermo Scientific™ BSA Silylation Reagent. This reagent reacts readily with a wide range of compounds, including alcohols, amines, carboxylic acids, phenols, steroids, biogenic amines, and alkaloids to form volatile, stable TMS derivatives for GC.
Recommended Use
Not recommended for use with carbohydrates or very low molecular weight compounds
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
General References:
  1. Tai, S.S., et al. (1994). The certification of morphine and codeine in a human urine standard reference material. J. of Toxicology 18, 7
  2. Welch, M.J., et al. (1993). Hair analysis for drugs of abuse. J. of Toxicology 17, 389
Specifications
For Use WithGC
Standard Concentration3 to 5%
HazardsCorrosive, Flammable
Item Description100 g
Storage2°C to 8°C
Quantity100 g
Standard ComponentsN-(Trimethylsilyl)acetamide
Unit SizeEach