Hydrocarbon Soluble Siliconizing Fluid
Hydrocarbon Soluble Siliconizing Fluid
Thermo Scientific™
Hydrocarbon Soluble Siliconizing Fluid
For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TS-42800120 mL
TS-42801480 mL
Catalog number TS-42800
Price (USD)
-
Request A Quote
Quantity:
120 mL
For siliconizing glass surfaces use Thermo Scientific™ Hydrocarbon Soluble Siliconizing Fluid. This short chain, clear polymeric silicone fluid consists primarily of dichlorooctamethyltetrasiloxane and is soluble in organic solvents. When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tiightly bonded film over the entire surface. It is excellent for modifying metals, certain plastics, ceramics and fiber optics.
Additional Product Information
Hydrocarbon Soluble Siliconizing Fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid.
The fluid is acidic only during application; after application, the surface is neutral.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithGC
HazardsCorrosive to Eyes/Skin/Respiratory Tract
Item Description120 mL
Quantity120 mL
Standard Components1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxane
Product TypeHydrocarbon Soluble Siliconizing Fluid
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection