Search Thermo Fisher Scientific
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent. It prevents the formation of multiple derivatives when enols are present during silylation, thereby ensuring optimal quantitation. The reagent is a 2% solution of methoxyamine–hydrogen chloride in pyridine.
General References: