Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
Thermo Scientific™
Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent.
Have Questions?
Catalog number TS-58220
Price (USD)/ Each
320.83
-
Add to cart
For Use With:
GC
Price (USD)/ Each
320.83
Add to cart
Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent
Catalog numberTS-58220
Price (USD)/ Each
320.83
-
Add to cart
Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent. Reaction times are fast (∼20 minutes). Derivatives are highly EC-sensitive, making them useful in low-level determinations of fatty acids. PFBBr has been applied in analyzing trace organics in asphalts and as a fingerprinting technique for identifying asphalt pollutants found in surface waters.
  • Fast reaction times for extraction alkylation technique: ∽20 minutes
  • Derivatives are highly EC-sensitive, making them useful in low-level determinations of fatty acids
  • Analysis of trace organics in asphalt
Recommended for:
Analysis of trace organics in asphalt
For Research Use Only. Not for use in diagnostic procedures.
Specifications
StorageRoom Temperature
For Use WithGC
HazardsCorrosive
Standard Componentsalpha-Bromo-2,3,4,5,6-pentafluorotoluene
Standard Concentration98 to 100%
Quantity5 g
Unit SizeEach