Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
TS-84769 | 100 g |
TS-84770 | 25 g |
Thermo Scientific™ HMDS (Hexamethyldisilazane) greatly extends the practical range of GC, improving chromatographic results in the silylation of sugars and related substances.
General References:
Share catalog number, name or link