TMSI Silylation Reagent
TMSI Silylation Reagent
Actual product may vary
Thermo Scientific™

TMSI Silylation Reagent

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TS-8862310 x 1 mL ampules
TS-8862525 g
2 Options
Catalog number TS-88623
Price (USD)/ Each
149.92
-
Add to cart
Quantity:
10 x 1 mL ampules
Price (USD)/ Each
149.92
Add to cart
TMSI Silylation Reagent
Catalog numberTS-88623
Price (USD)/ Each
149.92
-
Add to cart

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent. This silylation reagent reacts quickly with hydroxyls and carboxylic acids but not with amines, and is particularly well suited in multiderivatization schemes for compounds containing both hydroxyl and amine groups. TMSI may be used to derivatize alcohols, phenols, organic acids, hormones, glycols, nucleotides, and narcotics and is excellent for C1–C5 fatty acids in serum and urine.

  • Reacts quickly and smoothly with hydroxyls and carboxylic acids but not with amines
  • Especially useful in multiderivatization schemes for compounds containing both hydroxyl and amine groups
  • Used in the derivatization of alcohols, phenols, organic acids, steroids, hormones, glycols, nucleotides and narcotics
  • Excellent for C1 through C5 fatty acids in serum and urine
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithGC/MS
Standard Concentration98 to 100%
HazardsFlammable, Irritant
Item Description10 x 1 mL ampules
Storage2°C to 8°C
Quantity10 x 1 mL ampules
Standard Components1-(Trimethylsilyl)-1 H-imidazole
Unit SizeEach