Search Thermo Fisher Scientific
Accepts | Variety of plate types |
Description | F7 Industrial Robot |
Certifications/Compliance | RIA/CE |
Voltage | 110 V/230 V |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TS-F7000 also known as F7000 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Compact but mighty, the F7 robot has a small footprint, but unmatched power to handle sophisticated workflows. Its compact controller and integrated lab-oriented software provides reliable performance and increase speed of movement of a wide variety of labware. Track options provide further flexibility and the robot is able to access even the most challenging instruments due to the six-axis motion and wide range of available gripper fingers.
Features and Benefits
Options