Precision™ Circulating Water Baths
Precision™ Circulating Water Baths
Thermo Scientific™

Precision™ Circulating Water Baths

Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionCapacity (Metric)Electrical RequirementsModel
TSCIR1919L Circulating Water Bath19 LGlobal Voltage 100/115 V 200/230 V 50/60 HzPrecision CIR 19
TSCIR3535L Circulating Water Bath35 LGlobal Voltage 100/115 V 200/230 V 50/60 HzPrecision CIR 35
TSCIR8989L Circulating Water Bath89 LGlobal Voltage 100/115 V 200/230 V 50/60 HzPrecision CIR 89
Catalog number TSCIR19
Price (USD)
3,492.04
Online Exclusive
4,050.00
Save 557.96 (14%)
Each
Add to cart
Description:
19L Circulating Water Bath
Capacity (Metric):
19 L
Electrical Requirements:
Global Voltage 100/115 V 200/230 V 50/60 Hz
Model:
Precision CIR 19
Price (USD)
3,492.04
Online Exclusive
4,050.00
Save 557.96 (14%)
Each
Add to cart

Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.

  • Achieve enhanced temperature uniformity with perimeter-directed water flow
  • Easily clean and maintain bath with coil-free internal design
  • Optimize scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms
Not for use for IVD/MD in some countries.
Specifications
Amperage9.0-10.5 / 4.3-5.0 (120V/230V)
Capacity (English)5.0 gal.
Certifications/ComplianceUL, CE
CommunicationsDigital
Controller TypeDigital
Description19L Circulating Water Bath
Dimensions (D x W x H) Interior12 x 15.3 x 7.6 in. (305 x 387 x 192 mm)
Dimensions (L x W x H)15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
DisplayMonochrome LCD
Heating Capacity1200 w
Includes1 Stainless Steel Gable cover and Rubber Duck
Material304 Stainless Steel
ModelPrecision CIR 19
Temperature Presets4
Temperature Stability±0.1°C
Temperature Uniformity±0.05°C
Capacity (Metric)19 L
Electrical RequirementsGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Temperature RangeAmbient +5° to 100°C
Unit SizeEach

Share catalog number, name or link