Thermo Scientific™

CryoMed™ with OPC UA, General Purpose

Catalog number: TSCM34SA
Have Questions?
Thermo Scientific™

CryoMed™ with OPC UA, General Purpose

Catalog number: TSCM34SA
Have Questions?
For customers in cell and gene therapy and vaccine production, the CryoMed with OPC UA provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed features OPC UA serial communication (ethernet) capabilities, comes standard factory certifications, provides enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
 
Catalog Number
TSCM34SA
Unit Size
Each
Description
General purpose with printer, 1.2 cu. ft. (34 L)
Dimensions (D x W x H) Interior
13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Vial Capacity
<800
Price (USD)
Full specifications
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Electrical Requirements120V, 50-60Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose with printer, 1.2 cu. ft. (34 L)
Dimensions (D x W x H) Interior
13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Vial Capacity
<800
Price (USD)
Full specifications
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120V, 50-60Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose with printer, 1.2 cu. ft. (34 L)
Dimensions (D x W x H) Interior
13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Vial Capacity
<800
Price (USD)
Full specifications
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose with printer, 1.2 cu. ft. (34 L)
Dimensions (D x W x H) Interior
13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Vial Capacity
<800
Price (USD)
Full specifications
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230 V, 50-60 Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose with printer, 1.7 cu. ft. (48 L)
Dimensions (D x W x H) Interior
13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Vial Capacity
<1185
Price (USD)
Full specifications
DescriptionGeneral purpose with printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120V, 50-60Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose with printer, 1.7 cu. ft. (48 L)
Dimensions (D x W x H) Interior
13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Vial Capacity
<1185
Price (USD)
Full specifications
DescriptionGeneral purpose with printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/ComplianceCE
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose without printer, 1.7 cu. ft. (48 L)
Dimensions (D x W x H) Interior
13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Vial Capacity
<1185
Price (USD)
Full specifications
DescriptionGeneral purpose without printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120 V, 50-60 Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose without printer, 1.7 cu. ft. (48 L)
Dimensions (D x W x H) Interior
13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Vial Capacity
<1185
Price (USD)
Full specifications
DescriptionGeneral purpose without printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose with printer, 0.6 cu. ft. (17 L)
Dimensions (D x W x H) Interior
13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Vial Capacity
<380
Price (USD)
Full specifications
DescriptionGeneral purpose with printer, 0.6 cu. ft. (17 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<380
Weight (English)154 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
General purpose without printer, 0.6 cu. ft. (17 L)
Dimensions (D x W x H) Interior
13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
Dimensions (DxWxH) Exterior
24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Vial Capacity
<380
Price (USD)
Full specifications
DescriptionGeneral purpose without printer, 0.6 cu. ft. (17 L)
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120V, 50-60Hz
Vial Capacity<380
Weight (English)154 lb.
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionDimensions (D x W x H) InteriorDimensions (DxWxH) ExteriorVial CapacityPrice (USD)
TSCM34SAFull specifications
EachGeneral purpose with printer, 1.2 cu. ft. (34 L)13 x 13 x 12 in. (33 x 33 x 30.5 cm)24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)<800Request A Quote
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Electrical Requirements120V, 50-60Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
TSCM34FAFull specifications
EachGeneral purpose with printer, 1.2 cu. ft. (34 L)13 x 13 x 12 in. (33 x 33 x 30.5 cm)24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)<800Request A Quote
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120V, 50-60Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
TSCM34FVFull specifications
EachGeneral purpose with printer, 1.2 cu. ft. (34 L)13 x 13 x 12 in. (33 x 33 x 30.5 cm)24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)<800Request A Quote
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
TSCM34SVFull specifications
EachGeneral purpose with printer, 1.2 cu. ft. (34 L)13 x 13 x 12 in. (33 x 33 x 30.5 cm)24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)<800Request A Quote
DescriptionGeneral purpose with printer, 1.2 cu. ft. (34 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230 V, 50-60 Hz
Vial Capacity<800
Weight (English)174 lb.
Unit SizeEach
TSCM48FAFull specifications
EachGeneral purpose with printer, 1.7 cu. ft. (48 L)13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)<1185Request A Quote
DescriptionGeneral purpose with printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120V, 50-60Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
TSCM48FVFull specifications
EachGeneral purpose with printer, 1.7 cu. ft. (48 L)13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)<1185Request A Quote
DescriptionGeneral purpose with printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/ComplianceCE
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
TSCM48SAFull specifications
EachGeneral purpose without printer, 1.7 cu. ft. (48 L)13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)<1185Request A Quote
DescriptionGeneral purpose without printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120 V, 50-60 Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
TSCM48SVFull specifications
EachGeneral purpose without printer, 1.7 cu. ft. (48 L)13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)<1185Request A Quote
DescriptionGeneral purpose without printer, 1.7 cu. ft. (48 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<1185
Weight (English)191 lb.
Unit SizeEach
TSCM17FVFull specifications
EachGeneral purpose with printer, 0.6 cu. ft. (17 L)13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)<380Request A Quote
DescriptionGeneral purpose with printer, 0.6 cu. ft. (17 L)
TypeControlled-rate freezer
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/ComplianceCE
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeCEE 7/7
Electrical Requirements220-230V, 50-60Hz
Vial Capacity<380
Weight (English)154 lb.
Unit SizeEach
TSCM17SAFull specifications
EachGeneral purpose without printer, 0.6 cu. ft. (17 L)13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)<380Request A Quote
DescriptionGeneral purpose without printer, 0.6 cu. ft. (17 L)
Temperature Range-180° to +50 °C
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated steel
Certifications/CompliancecULus
DisplayTouchscreen user interface
WarrantyWarranty varies by region. Contact your local sales representative for more information
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 7 x 12 in. (33 x 17.8 x 30.5 cm)
Data OutputsUSB and OPC UA (ethernet)
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
ConnectivityThermo Fisher Connect compatible
Amperage12 A
Plug TypeNEMA 5-15 P
Electrical Requirements120V, 50-60Hz
Vial Capacity<380
Weight (English)154 lb.
Unit SizeEach
Showing 10 of 12Show more

User-friendly operation with enhanced data traceability

  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • OPC UA (ethernet) serial communication protocol
  • Factory-issued certificates
  • Six pre-set freezing profiles and space for up to 14 user-defined, “custom” freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • “Run last” feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation

Real-time run monitoring for sample protection

  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing “real time” responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer

Reliable temperature performance for high throughput usage

  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish

Warranty varies by region. Check with your local representative for more information.

Services and Support (optional)
When you invest in Thermo Scientific laboratory equipment, you not only benefit from top-quality product performance, but you can also enjoy peace of mind and maximum uptime with world-class service solutions provided by Unity Lab Services.
 
Service plans* – Maximize uptime, maintain peak performance, and extend the life of your equipment with a service plan that includes annual preventive maintenance, digital remote resolution, and fast onsite response times.

Catalog numbers:

  • Factory Warranty Period
    • Total Care Warranty TCWTYSCRYO
  • Post Warranty Period
    • Extended Warranty EXTWSCRYO
    • Tech Direct Service Plan TEDSCRYO
    • Total Care Service Plan TLCSCRYO

Compliance Services** – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.  
Catalog numbers:

Qualification service

  • On-site IQ/OQ service IOQP003507
  • On-site IQ/OQ + temperature mapping IOTQP003507
  • On-site temperature mapping service TEMPMAP

*Available in 23 countries. Please indicate your interest in the “Request a quote” form and we will come back to you on availability.
 
**Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
 

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text