Thermo Scientific™
TSQ Quantis™ Triple Quadrupole Mass Spectrometer
Catalog number: TSQ02-10001
Have Questions?
Thermo Scientific™
TSQ Quantis™ Triple Quadrupole Mass Spectrometer
Catalog number: TSQ02-10001
Have Questions?
Easily answer critical analytical challenges during targeted quantitation workflows in your laboratory. With its segmented quadrupoles, faster rod drivers, and improved electron multipliers, the Thermo Scientific™ TSQ Quantis™ Triple Quadrupole Mass Spectrometer delivers unprecedented robustness with best-in-class sensitivity for everyday use and for every user.
Catalog Number
TSQ02-10001
Unit Size
Each
Description
TSQ Quantis triple-stage quadrupole mass spectrometer
Price (USD)
Full specifications
TypeTriple Quadrupole LC-MS
Mass Range5 to 3000 m/z
DescriptionTSQ Quantis triple-stage quadrupole mass spectrometer
For Use With (Application)Pharmaceutical, Environmental, Food Safety, Clinical Research, and Forensic Toxicology
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
TSQ02-10001Full specifications
EachTSQ Quantis triple-stage quadrupole mass spectrometerRequest A Quote
TypeTriple Quadrupole LC-MS
Mass Range5 to 3000 m/z
DescriptionTSQ Quantis triple-stage quadrupole mass spectrometer
For Use With (Application)Pharmaceutical, Environmental, Food Safety, Clinical Research, and Forensic Toxicology
Unit SizeEach
Showing 1 of 1
The TSQ Quantis MS seamlessly integrates with industry-leading quantitation software, HPLCs and UHPLCs, and sample preparation solutions; it also offers enhanced efficiency during targeted quantitation workflows for a wide array of molecular species. Other benefits include:
  • Superb sensitivity that generates data for all molecular species, even in complex matrices.
  • Outstanding robustness that enables increased confidence in data while prolonging instrument uptime.
  • Reliability and reproducibility improve data quality for every run and every sample.
  • Close integration with application-specific software ensures increased productivity across all application markets.
  • Simplicity and ease-of-use allow users of all expertise levels to acquire high quality data with improved confidence in results.