Thermo Scientific™

TSQ Altis™ Triple Quadrupole Mass Spectrometer

Catalog number: TSQ02-10002
Have Questions?
Thermo Scientific™

TSQ Altis™ Triple Quadrupole Mass Spectrometer

Catalog number: TSQ02-10002
Have Questions?

Address the most stringent analytical challenges for targeted quantitation workflows with the Thermo Scientific™ TSQ Altis™ Triple Quadrupole Mass Spectrometer. With its improved Active Ion Management (AIM™) technology, segmented quadrupoles, novel electron multipliers, and enhanced ion transmission tubes, the TSQ Altis MS helps you achieve unprecedented experimental sensitivity for all molecular species in complex matrices without sacrificing robustness.

 
Catalog Number
TSQ02-10002
Unit Size
Each
Description
TSQ Altis triple-stage quadrupole mass spectrometer
Price (USD)
Full specifications
TypeTriple Quadrupole LC-MS
Mass Range5 to 2000 m/z
DescriptionTSQ Altis triple-stage quadrupole mass spectrometer
For Use With (Application)Pharmaceutical, Environmental, Food Safety, Clinical Research, and Forensic Toxicology
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
TSQ02-10002Full specifications
EachTSQ Altis triple-stage quadrupole mass spectrometerRequest A Quote
TypeTriple Quadrupole LC-MS
Mass Range5 to 2000 m/z
DescriptionTSQ Altis triple-stage quadrupole mass spectrometer
For Use With (Application)Pharmaceutical, Environmental, Food Safety, Clinical Research, and Forensic Toxicology
Unit SizeEach
Showing 1 of 1
Sensitivity with Robustness: No Compromise

The TSQ Altis Triple Quadrupole Mass Spectrometer, together with its industry-leading quantitation software, UHPLC system, and superior sample preparation kits, helps researchers address critical analytical challenges using robust, reliable, and sensitive targeted quantitation. The main benefits of the TSQ Altis MS include:

  • Ultimate sensitivity for all molecular types in matrices ranging from simple to complex.
  • Outstanding instrument robustness enables increased confidence in generated data with no loss of instrument uptime.
  • Ultrafast selected reaction monitoring (SRM) increases the amount of molecular quantifications in less time.
  • Close integration with application-specific software results in increased productivity across all application markets.
  • Simplicity and ease-of-use allow users of all expertise levels to acquire high quality results with increased confidence.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text