Precision™ Shaking Water Baths
Precision™ Shaking Water Baths
Thermo Scientific™

Precision™ Shaking Water Baths

Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelDescriptionCapacity (Metric)Electrical Requirements
TSSWB27Precision SWB 2727L Shaking Water Bath27 LGlobal Voltage 100/115 V 200/230 V 50/60 Hz
TSSWB15Precision SWB 1515L Shaking Water Bath15 LGlobal Voltage 100/115 V 200/230 V 50/60 Hz
TSSWB15SPrecision SWB 15S15L Shallow Shaking Water Bath15 LGlobal Voltage 100/115 V 200/230 V 50/60 Hz
TSDUB15Precision DUB 1515L Dubnoff Shaking Water Bath15 LGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Catalog number TSSWB27
Price (EUR)
3.600,00
Each
-
Add to cart
Model:
Precision SWB 27
Description:
27L Shaking Water Bath
Capacity (Metric):
27 L
Electrical Requirements:
Global Voltage 100/115 V 200/230 V 50/60 Hz
Price (EUR)
3.600,00
Each
Add to cart
Ask our AI about this Product

Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere

  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor
  • Warranty varies by country, please contact us for details
Not for use for IVD/MD in some countries.
Specifications
Amperage11.2-13.1 / 5.4-6.3 (120V/230V)
Certifications/ComplianceUL, CE
Chamber Material304 Stainless Steel
Controller TypeDigital
Description27L Shaking Water Bath
Dimensions (L x W x H)15.5 x 36.9 x 9.8 in. (394 x 938 x 249 mm)
DisplayMonochrome LCD
Heating Capacity1500 w
IncludesStainless Steel Gable Cover and Rubber Duck
MaterialEpoxy Powder-coated Cold Rolled Steel
ModelPrecision SWB 27
Plug TypeN5-20. (Requires a 20A receptacle.)
Temperature Presets4
Temperature Stability±0.1°C
Temperature Uniformity±0.05°C
Capacity (Metric)27 L
Electrical RequirementsGlobal Voltage 100/115 V 200/230 V 50/60 Hz
Temperature RangeAmbient +5° to 100°C
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Share catalog number, name or link

1x1 image pixel for data collection